accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOOOOH2OH2OH2OH2OH2OH2OH2OH2OPrPr

CAS RN 14948-62-0

IUPAC Name: dipraseodymium(3+) octahydrate tricarbonate
Synonyms: dipraseodymium(3+) octahydrate tricarbonate
Molecular Weight (g/mol): 605.96
Molecular Formula: C3H16O17Pr2
InChi Key: SEHQEMTYQGRHIC-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Pr+3].[Pr+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Physical Form 
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)