accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ZrNNOOOOOOO

CAS RN 14985-18-3

IUPAC Name: oxozirconiumbis(ylium) hydrate dinitrate
Synonyms: oxozirconiumbis(ylium) hydrate dinitrate
Molecular Weight (g/mol): 249.25
Molecular Formula: H2N2O8Zr
InChi Key: BGMQNYCWALSARE-UHFFFAOYSA-N
SMILES: O.O=[Zr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)