accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1499-10-1

IUPAC Name: 9,10-diphenylanthracene
Synonyms: 9,10-diphenylanthracene
Molecular Weight (g/mol): 330.43
Molecular Formula: C26H18
InChi Key: FCNCGHJSNVOIKE-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results