accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 149947-16-0

BrFBr

CAS RN 149947-16-0

IUPAC Name: 1-bromo-3-(bromomethyl)-2-fluorobenzene
Synonyms: 1-bromo-3-(bromomethyl)-2-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: MGVHFTWDCYIBDO-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC=C1Br
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)