accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 149949-29-1

IUPAC Name: 2-chloro-6-fluoro-3-methoxybenzaldehyde
Synonyms: 2-chloro-6-fluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: WRJUPJLEUPNEBO-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C(C=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results