accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14996-61-3

IrClClCl

CAS RN 14996-61-3

IUPAC Name: trichloroiridium hydrate
Synonyms: trichloroiridium hydrate
Molecular Weight (g/mol): 316.58
Molecular Formula: Cl3H2IrO
InChi Key: MJRFDVWKTFJAPF-UHFFFAOYSA-K
SMILES: O.Cl[Ir](Cl)Cl
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results