accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14999-52-1

IUPAC Name: disodium 9-{5-[(hydrogen phosphonatooxy)methyl]-4-hydroxyoxolan-2-yl}-9H-purin-6-olate
Synonyms: disodium 9-{5-[(hydrogen phosphonatooxy)methyl]-4-hydroxyoxolan-2-yl}purin-6-olate
Molecular Weight (g/mol): 376.17
Molecular Formula: C10H11N4Na2O7P
InChi Key: VDPLPYMIJCLJEF-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC1CC(OC1COP(O)([O-])=O)N1C=NC2=C([O-])N=CN=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)