accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 150-30-1

IUPAC Name: 2-amino-3-phenylpropanoic acid
Synonyms: phenylalanin sabiden phenylalanine DL-form DL-3-phenylalanine alanine, phenyl-, DL- (8CI) alanine, phenyl-, D- alanine, phenyl-, dl- fenilalanina
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (13)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results