accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 150-60-7

IUPAC Name: [(benzyldisulfanyl)methyl]benzene
Synonyms: disulfide, bis(phenylmethyl)
Molecular Weight (g/mol): 246.39
Molecular Formula: C14H14S2
InChi Key: GVPWHKZIJBODOX-UHFFFAOYSA-N
SMILES: C(SSCC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)