accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 150-61-8

NHHN

CAS RN 150-61-8

IUPAC Name: N1,N2-diphenylethane-1,2-diamine
Synonyms: diphenylethylenediamine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: NOUUUQMKVOUUNR-UHFFFAOYSA-N
SMILES: C(CNC1=CC=CC=C1)NC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results