accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3COHOO

CAS RN 150-83-4

IUPAC Name: sodium 3-hydroxybutanoate
Synonyms: sodium 3-hydroxybutyrate
Molecular Weight (g/mol): 126.09
Molecular Formula: C4H7NaO3
InChi Key: NBPUSGBJDWCHKC-UHFFFAOYNA-M
SMILES: [Na+].CC(O)CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results