accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NPdNClCl

CAS RN 15020-99-2

IUPAC Name: dichloropalladium; ethane-1,2-diamine
Synonyms: ethylenediamine; palladium chloride
Molecular Weight (g/mol): 237.42
Molecular Formula: C2H8Cl2N2Pd
InChi Key: CAYKJANQVKIYPJ-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.NCCN
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results