accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBHON

CAS RN 150255-96-2

IUPAC Name: (3-cyanophenyl)boronic acid
Synonyms: 3-cyanophenylboronic acid
Molecular Weight (g/mol): 146.94
Molecular Formula: C7H6BNO2
InChi Key: XDBHWPLGGBLUHH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results