CAS RN 1504-55-8
IUPAC Name:
(3E)-4-phenylbut-3-en-2-ol
Synonyms:
methylcinnamyl alcohol
Molecular Weight (g/mol):
148.21
Molecular Formula:
C10H12O
InChi Key:
ZIJWGEHOVHJHKB-BQYQJAHWNA-N
SMILES:
CC(O)\C=C\C1=CC=CC=C1
1
–
1
of
1
results
1
trans-2-Methyl-3-phenyl-2-propen-1-ol, 95%
CAS: 1504-55-8 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.2 InChI Key: LLNAMUJRIZIXHF-VQHVLOKHSA-N Synonym: 2-methyl-3-phenyl-2-propen-1-ol,beta-methylcinnamyl alcohol,2-propen-1-ol, 2-methyl-3-phenyl,alpha-methylcinnamyl alcohol,methyl cinnamic alcohol,alpha-methylcinnamic alcohol,alpha methyl cinnamic alcohol,3-phenyl-2-methyl-propen-2-ol-1,e-2-methyl-3-phenyl-2-propen-1-ol,2e-2-methyl-3-phenylprop-2-en-1-ol PubChem CID: 6393846 IUPAC Name: (E)-2-methyl-3-phenylprop-2-en-1-ol SMILES: CC(=CC1=CC=CC=C1)CO