accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15049-67-9

CH3CH3OCH3CH3OCH3CH3OBi

CAS RN 15049-67-9

IUPAC Name: bismuth(3+) tris(propan-2-olate)
Synonyms: bismuth(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 386.24
Molecular Formula: C9H21BiO3
InChi Key: KNPRLIQQQKEOJN-UHFFFAOYSA-N
SMILES: [Bi+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)