CAS RN 150517-76-3
IUPAC Name:
1-(2,4-dichloro-6-methylphenyl)methanamine
Synonyms:
1-(2,4-dichloro-6-methylphenyl)methanamine
Molecular Weight (g/mol):
190.07
Molecular Formula:
C8H9Cl2N
InChi Key:
QISTWSMZJRTDCS-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=CC(Cl)=C1CN
1
–
1
of
1
results
1
2,4-Dichloro-6-methylbenzylamine, 98+%, Thermo Scientific™
CAS: 150517-76-3 Molecular Formula: C8H9Cl2N Molecular Weight (g/mol): 190.07 MDL Number: MFCD00052913 InChI Key: QISTWSMZJRTDCS-UHFFFAOYSA-N Synonym: 2,4-dichloro-6-methylbenzylamine,2,4-dichloro-6-methylphenyl methanamine,2,4-dichloro-6-methylphenyl methylamine,benzenemethanamine,2,4-dichloro-6-methyl,benzenemethanamine, 2,4-dichloro-6-methyl,2,4-dichloro-6-methylbenzyl amine hydrochloride,4,6-dichloro-2-methylbenzylamine,acmc-1c3je,2-aminomethyl-3,5-dichlorotoluene,1-2,4-dichloro-6-methylphenyl methanamine PubChem CID: 2800971 SMILES: CC1=CC(Cl)=CC(Cl)=C1CN