accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONNClNNCl

CAS RN 150728-13-5

IUPAC Name: 4,6-dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
Synonyms: 4,6-dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
Molecular Weight (g/mol): 349.17
Molecular Formula: C15H10Cl2N4O2
InChi Key: IZGOBGVYADHVKH-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC1=C(Cl)N=C(N=C1Cl)C1=NC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results