accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 150812-21-8

NHNH2NOOF

CAS RN 150812-21-8

IUPAC Name: N1-[(4-fluorophenyl)methyl]-3-nitrobenzene-1,4-diamine
Synonyms: N1-[(4-fluorophenyl)methyl]-3-nitrobenzene-1,4-diamine
Molecular Weight (g/mol): 261.26
Molecular Formula: C13H12FN3O2
InChi Key: XTDZJOIEYRRRGJ-UHFFFAOYSA-N
SMILES: NC1=C(C=C(NCC2=CC=C(F)C=C2)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results