accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15098-87-0

H2OH2OH2OFFFAl

CAS RN 15098-87-0

IUPAC Name: aluminium(3+) trihydrate trifluoride
Synonyms: aluminium(3+) trihydrate trifluoride
Molecular Weight (g/mol): 138.02
Molecular Formula: AlF3H6O3
InChi Key: BKLZIAYVINRQEJ-UHFFFAOYSA-K
SMILES: O.O.O.[F-].[F-].[F-].[Al+3]
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)