accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 151-10-0

IUPAC Name: 1,3-dimethoxybenzene
Synonyms: M-dimethoxybenzene
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: DPZNOMCNRMUKPS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results