CAS RN 151-21-3
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate,SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium lauryl sulfate,SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate (SDS), ≥95%
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eur.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: sodium dodecyl sulfate,sodium lauryl sulfate,sodium dodecylsulfate,sodium lauryl sulphate,sodium dodecyl sulphate,dodecyl sulfate, sodium salt,anticerumen,gardinol,neutrazyme,duponal PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecylsulfate, 10% Solution, Ultrapure
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate, 99+%, for Electrophoresis, Fisher Chemical™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: sodium dodecyl sulfate,sodium lauryl sulfate,sodium dodecylsulfate,sodium lauryl sulphate,sodium dodecyl sulphate,dodecyl sulfate, sodium salt,anticerumen,gardinol,neutrazyme,duponal PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99%, for biochemistry
CAS: 151-21-3 Molecular Formula: C12H25NaO4S
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Sodium n-dodecyl sulfate, 97%, for electrophoresis
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Dodecyl sulfate, sodium salt, 85%, pure
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.38 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Dodecyl Sodium Sulfate,SDS,Sodium lauryl sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol