accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 151169-74-3

IUPAC Name: (2,3-dichlorophenyl)boronic acid
Synonyms: 2,3-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: TYIKXPOMOYDGCS-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C(Cl)=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results