accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFBrH2C

CAS RN 1514-82-5

IUPAC Name: 2-bromo-3,3,3-trifluoroprop-1-ene
Synonyms: 2-bromo-3,3,3-trifluoroprop-1-ene
Molecular Weight (g/mol): 174.95
Molecular Formula: C3H2BrF3
InChi Key: QKBKGNDTLQFSEU-UHFFFAOYSA-N
SMILES: FC(F)(F)C(Br)=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)