CAS RN 1515-95-3
IUPAC Name:
1-ethyl-4-methoxybenzene
Synonyms:
benzene, 1-ethyl-4-methoxy-
Molecular Weight (g/mol):
136.19
Molecular Formula:
C9H12O
InChi Key:
HDNRAPAFJLXKBV-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(OC)C=C1
1
–
1
of
1
results
1
4-Ethylanisole, 98+%
CAS: 1515-95-3 Molecular Formula: C9H12O Molecular Weight (g/mol): 136.194 MDL Number: MFCD00015176 InChI Key: HDNRAPAFJLXKBV-UHFFFAOYSA-N Synonym: 4-ethylanisole,p-ethylanisole,benzene, 1-ethyl-4-methoxy,anisole, p-ethyl,p-ethylanisol,unii-i71p5qg20s,4-ethyl-1-methoxybenzene,4-ethyl anisole,4-ethylmethoxybenzene,acmc-1btzv PubChem CID: 73690 IUPAC Name: 1-ethyl-4-methoxybenzene SMILES: CCC1=CC=C(C=C1)OC