accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOHBrOCH3(E/Z)

CAS RN 151539-52-5

IUPAC Name: 3-(2-bromo-4,5-dimethoxyphenyl)prop-2-enoic acid
Synonyms: 3-(2-bromo-4,5-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 287.11
Molecular Formula: C11H11BrO4
InChi Key: MSJCWIFRXCLULX-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=CC(O)=O)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results