CAS RN 151720-43-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oxamflatin, TRC
CAS: 151720-43-3 Molecular Formula: C17 H14 N2 O4 S Molecular Weight (g/mol): 342.37 IUPAC Name: (E)-5-[3-(benzenesulfonamido)phenyl]-N-hydroxypent-2-en-4-ynamide SMILES: ONC(=O)\C=C\C#Cc1cccc(NS(=O)(=O)c2ccccc2)c1