CAS RN 15178-63-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(1-Oxyl-2,2,6,6-tetramethyl-4-piperidinyl)maleimide, TRC
CAS: 15178-63-9 Molecular Formula: C13 H19 N2 O3 Molecular Weight (g/mol): 251.3 IUPAC Name: 1-(1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)pyrrole-2,5-dione SMILES: CC1(C)CC(CC(C)(C)N1[O])N2C(=O)C=CC2=O