accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3H3CCH3H3CCH3

CAS RN 15181-11-0

IUPAC Name: 1,3-di-tert-butyl-5-methylbenzene
Synonyms: 1,3-di-tert-butyl-5-methylbenzene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: WIXDSJRJFDWTNY-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results