CAS RN 1519-70-6
IUPAC Name:
[4-(carbamothioylamino)phenyl]thiourea
Synonyms:
4-(carbamothioylamino)phenylthiourea
Molecular Weight (g/mol):
226.32
Molecular Formula:
C8H10N4S2
InChi Key:
AMNPXXIGUOKIPP-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=CC=C(NC(N)=S)C=C1
1
–
1
of
1
results
1
N,N″-1,4-Phenylenebis(thiourea), 98+%, Thermo Scientific™
CAS: 1519-70-6 Molecular Formula: C8H10N4S2 Molecular Weight (g/mol): 226.32 MDL Number: MFCD00022169 InChI Key: AMNPXXIGUOKIPP-UHFFFAOYSA-N Synonym: 1,4-phenylenebis 2-thiourea,urea, 1,1'-p-phenylenebis 2-thio,1,1'-p-phenylenebis 2-thiourea,thiourea, n,n-1,4-phenylenebis,4-carbamothioylamino phenylthiourea,4-carbamothioylamino phenyl thiourea,1,1'-benzene-1,4-diylbis thiourea,1,4-phenylenebis,phenylenethiourea,acmc-20ap1t PubChem CID: 684351 IUPAC Name: [4-(carbamothioylamino)phenyl]thiourea SMILES: NC(=S)NC1=CC=C(NC(N)=S)C=C1