accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OPSOH3COH3C

CAS RN 152-18-1

IUPAC Name: O,O,O-trimethyl phosphorothioate
Synonyms: O,O,O-trimethyl phosphorothioate
Molecular Weight (g/mol): 156.14
Molecular Formula: C3H9O3PS
InChi Key: XWSLYQXUTWUIKM-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)