accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15205-27-3

IUPAC Name: [(2H-1,3-benzodioxol-5-yl)methyl](methyl)azanium
Synonyms: (2H-1,3-benzodioxol-5-ylmethyl)(methyl)azanium
Molecular Weight (g/mol): 166.2
Molecular Formula: C9H12NO2
InChi Key: CEPGPPSMCRKGFJ-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=C2OCOC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results