accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1521-51-3

IUPAC Name: 3-bromocyclohex-1-ene
Synonyms: 3-bromocyclohex-1-ene
Molecular Weight (g/mol): 161.04
Molecular Formula: C6H9Br
InChi Key: AJKDUJRRWLQXHM-UHFFFAOYNA-N
SMILES: BrC1CCCC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)