accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 152120-55-3

OONHNOONN(E/Z)

CAS RN 152120-55-3

IUPAC Name: benzyl N-({[(benzyloxy)carbonyl]imino}(1H-pyrazol-1-yl)methyl)carbamate
Synonyms: benzyl N-({[(benzyloxy)carbonyl]imino}(pyrazol-1-yl)methyl)carbamate
Molecular Weight (g/mol): 378.39
Molecular Formula: C20H18N4O4
InChi Key: NRBUVVTTYMTSKM-UHFFFAOYSA-N
SMILES: O=C(NC(=NC(=O)OCC1=CC=CC=C1)N1C=CC=N1)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results