accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1522-13-0

IUPAC Name: triphenylprop-2-yn-1-ol
Synonyms: triphenylprop-2-yn-1-ol
Molecular Weight (g/mol): 284.36
Molecular Formula: C21H16O
InChi Key: VWRQCJRTHKUVNF-UHFFFAOYSA-N
SMILES: OC(C#CC1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results