accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KCH3OOOSOOO

CAS RN 15220-11-8

IUPAC Name: potassium 4-methyl-2-oxo-2H-chromen-7-yl sulfate
Synonyms: potassium 4-methyl-2-oxochromen-7-yl sulfate
Molecular Weight (g/mol): 294.32
Molecular Formula: C10H7KO6S
InChi Key: CSOCSPXOODWGLJ-UHFFFAOYSA-M
SMILES: [K+].CC1=CC(=O)OC2=CC(OS([O-])(=O)=O)=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results