accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBr

CAS RN 15231-91-1

IUPAC Name: 6-bromonaphthalen-2-ol
Synonyms: 6-bromo-2-naphthol bromo-6 naphtol-2 C10H7BrO
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: YLDFTMJPQJXGSS-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=C(Br)C=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)