accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CoII

CAS RN 15238-00-3

IUPAC Name: λ²-cobalt(2+) diiodide
Synonyms: cobalt(II) iodide
Molecular Weight (g/mol): 312.74
Molecular Formula: CoI2
InChi Key: AVWLPUQJODERGA-UHFFFAOYSA-L
SMILES: [Co++].[I-].[I-]
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results