accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15244-38-9

CrOSOOOOSOOOOSOOOCr

CAS RN 15244-38-9

IUPAC Name: dichromium(3+) hydrate trisulfate
Synonyms: dichromium(3+) hydrate trisulfate
Molecular Weight (g/mol): 410.18
Molecular Formula: Cr2H2O13S3
InChi Key: KVIYQADEXNCDCB-UHFFFAOYSA-H
SMILES: O.[Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results