accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNHSO

CAS RN 15264-63-8

IUPAC Name: 5-(pyridin-4-yl)-2,3-dihydro-1,3,4-oxadiazole-2-thione
Synonyms: 5-(pyridin-4-yl)-3H-1,3,4-oxadiazole-2-thione
Molecular Weight (g/mol): 179.2
Molecular Formula: C7H5N3OS
InChi Key: TXCXZVFDWQYTIC-UHFFFAOYSA-N
SMILES: S=C1NN=C(O1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results