accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1527-91-9

IUPAC Name: [phenyl(phenylamino)methylidene]azanium
Synonyms: [phenyl(phenylamino)methylidene]azanium
Molecular Weight (g/mol): 197.26
Molecular Formula: C13H13N2
InChi Key: MPYOKHFSBKUKPQ-UHFFFAOYSA-O
SMILES: [NH2+]=C(NC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)