accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1529-48-2

ClHGeH2ClHCH3H3C

CAS RN 1529-48-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 177.63
Molecular Formula: C2H10Cl2Ge
InChi Key: PDIYLVAYJQRPJM-UHFFFAOYSA-N
SMILES: Cl.Cl.C[GeH2]C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)