accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1531-77-7

IUPAC Name: 9-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Synonyms: 9-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Molecular Weight (g/mol): 226.29
Molecular Formula: C14H10OS
InChi Key: JGJDEWXZEIHBNW-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2CSC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results