accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15321-51-4

CHOCHOCHOCOCOCOCOCOCOFeFe

CAS RN 15321-51-4

IUPAC Name: iron(3+) hexakis(methanidylidyneoxidanium) tris(oxomethanide) iron
Synonyms: diiron nonacarbonyl
Molecular Weight (g/mol): 366.8
Molecular Formula: C9H3Fe2O9
InChi Key: CGJSNKHFIBINMR-UHFFFAOYSA-N
SMILES: [Fe].[Fe+3].[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)