accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15329-69-8

OHOOHN(Z)

CAS RN 15329-69-8

IUPAC Name: (2Z)-3-(benzylcarbamoyl)prop-2-enoic acid
Synonyms: (2Z)-3-(benzylcarbamoyl)prop-2-enoic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: BHWGQIYJCMMSNM-SREVYHEPSA-N
SMILES: OC(=O)\C=C/C(=O)NCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results