accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PNOH(E/Z)

CAS RN 153358-05-5

IUPAC Name: N-{[2-(diphenylphosphanyl)phenyl]methylidene}hydroxylamine
Synonyms: N-{[2-(diphenylphosphanyl)phenyl]methylidene}hydroxylamine
Molecular Weight (g/mol): 305.32
Molecular Formula: C19H16NOP
InChi Key: XHIVESUSSLEMGJ-UHFFFAOYSA-N
SMILES: ON=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results