accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

RhClClClClClClNHHHHNHHHHNHHHH

CAS RN 15336-18-2

IUPAC Name: rhodium(3+) triammonium hexachloride
Synonyms: rhodium(3+) triammonium hexachloride
Molecular Weight (g/mol): 369.72
Molecular Formula: Cl6H12N3Rh
InChi Key: OJNYBYILSUWBHM-UHFFFAOYSA-K
SMILES: [NH4+].[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Rh+3]
Grade 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)