accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOONOO

CAS RN 15341-08-9

IUPAC Name: (5-nitro-2H-1,3-benzodioxol-4-yl)methanol
Synonyms: (5-nitro-2H-1,3-benzodioxol-4-yl)methanol
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: LJXBRPHBDWIHPR-UHFFFAOYSA-N
SMILES: OCC1=C2OCOC2=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)