accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOFF

CAS RN 1535-65-5

IUPAC Name: difluoromethanesulfonylbenzene
Synonyms: difluoromethanesulfonylbenzene
Molecular Weight (g/mol): 192.18
Molecular Formula: C7H6F2O2S
InChi Key: LRHDNAVPELLXDL-UHFFFAOYSA-N
SMILES: FC(F)S(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results