accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OFFF

CAS RN 1535-75-7

IUPAC Name: 2-(trifluoromethoxy)aniline
Synonyms: 2-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: ZFCOUBUSGHLCDT-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results