CAS RN 153559-76-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
LG 100268, TRC
CAS: 153559-76-3 Molecular Formula: C24 H29 N O2 Molecular Weight (g/mol): 363.49 IUPAC Name: 6-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)cyclopropyl]pyridine-3-carboxylic acid SMILES: Cc1cc2c(cc1C3(CC3)c4ccc(cn4)C(=O)O)C(C)(C)CCC2(C)C
2
R&D Systems™ LG 100268
A Tocris product, part of R&D Systems. Potent and selective RXR agonist; also activates RXR/PPARγ heterodimers